missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molidustat, MedChemExpress
Description
Molidustat (BAY 85-3934) is a novel inhibitor of hypoxia-inducible factor prolyl hydroxylase (HIF-PH) with mean IC50 values of 480 nM for PHD1, 280 nM for PHD2, and 450 nM for PHD3.
Specifications
Specifications
| Chemical Name or Material | Molidustat |
| Physical Form | Solid |
| CAS | 1154028-82-6 |
| Color | Off-White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C13H14N8O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | BAY 85-3934 |
| Solubility Information | DMSO : 5 mg/mL (15.91 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?