missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum solution for ICP/MS 10mg/L HNO3 matrix, Fisher Chemical™
BIOPHARMA

Product Code. 12957383 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Unit Size:
100mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12957383 100 mL 100mL
12947383 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12957383 Supplier Fisher Chemical Supplier No. S 634 100
  • ONLINE EXCLUSIVE PRICE
    58.90 €
    49.97 € / 100mL
    Save 8.93 €
    15% Off
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Packaging Bottle
Concentration 10 mg/L
Solvent or Matrix Water
Quantity 100 mL
Chemical Name or Material Molybdenum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.