Learn More
Monkeypox Virus A35 Antibody (0026), DyLight 488 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18812G
Product Code. 18568057
  • 625.00 € / 0.10mL
Description
Monkeypox Virus A35 Monoclonal antibody specifically detects Monkeypox Virus A35 in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monkeypox Virus A35
Monoclonal
DyLight 488
Mouse
Protein A purified
RUO
Virus
Purified
Western Blot, ELISA
0026
50mM Sodium Borate
This product is a recombinant monoclonal antibody made to recombinant Monkeypox Virus A35 and expressed from HEK293 cells.
0.1 mL
Primary
Store at 4°C in the dark.
IgG1