missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkeypox Virus B6R Recombinant Rabbit Monoclonal Antibody (004)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA560388
This item is not returnable.
View return policy
Description
Monkeypox Virus B6R Recombinant Monoclonal Antibody for Western Blot, ELISA
The monkeypox virus (MPXV) is the causative agent of the monkeypox disease. As a double-stranded DNA virus member of the genus Orthopoxvirus and the family Poxviridae, MPXV is similar to other pathogens of this group including variola (the causative agent of smallpox), vaccinia, and the cowpox virus. MPXV A29L, M1R and B6R, the orthologous to A27L, L1R and B5R VACV antigens respectively, are thought to be able to elicit a protective immune response against lethal MPXV challenge.
Specifications
| Monkeypox Virus B6R | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Rabbit | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 004 | |
| Unconjugated | |
| Monkeypox B6R | |
| Recombinant Monkeypox Virus (MPXV) B6R protein (Tyr18-His279). | |
| 20 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction