missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Monkeypox Virus B6R Recombinant Rabbit Monoclonal Antibody (006)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

159.00 € - 429.00 €

Specifications

Antigen Monkeypox Virus B6R
Clone 006
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30536085
GREENER_CHOICE
View Documents
Invitrogen™
MA560389
100 μL
429.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30534429
View Documents
Invitrogen™
MA560390
20 μL
159.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Monkeypox Virus B6R Recombinant Monoclonal Antibody for Western Blot, ELISA

The monkeypox virus (MPXV) is the causative agent of the monkeypox disease. As a double-stranded DNA virus member of the genus Orthopoxvirus and the family Poxviridae, MPXV is similar to other pathogens of this group including variola (the causative agent of smallpox), vaccinia, and the cowpox virus. MPXV A29L, M1R and B6R, the orthologous to A27L, L1R and B5R VACV antigens respectively, are thought to be able to elicit a protective immune response against lethal MPXV challenge.
Specifications

Specifications

Monkeypox Virus B6R
1 mg/mL
ELISA, Western Blot
Unconjugated
Rabbit
RUO
PBS with no preservative
Recombinant Monkeypox Virus (MPXV) B6R protein (Tyr18-His279).
Antibody
006
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Virus
Monkeypox B6R
Primary
Protein A
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.