missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Monkeypox Virus E8L Antibody (15), Novus Biologicals™
Mouse Monoclonal Antibody
195.00 € - 396.00 €
Specifications
| Antigen | Monkeypox Virus E8L |
|---|---|
| Clone | 15 |
| Dilution | Western Blot 1:2000-1:10000, ELISA 1:1000-1:2000, Sandwich ELISA Detection 1:1000-1:10000, Lateral Flow Assay 1-2 mg/mL |
| Applications | ELISA, Western Blot, Sandwich ELISA Detection, Lateral Flow |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043007
|
Novus Biologicals™
NBP3-30752-100ul |
100 μL |
396.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30042654
|
Novus Biologicals™
NBP3-30752-20ul |
20 μL |
195.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Monkeypox Virus E8L Monoclonal antibody specifically detects Monkeypox Virus E8L in Virus samples. It is validated for ELISA,Western Blot,Sandwich ELISA Detection,Lateral FlowSpecifications
| Monkeypox Virus E8L | |
| Western Blot 1:2000-1:10000, ELISA 1:1000-1:2000, Sandwich ELISA Detection 1:1000-1:10000, Lateral Flow Assay 1-2 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 μm filtered solution in PBS | |
| IgG1 | |
| Protein A purified |
| 15 | |
| ELISA, Western Blot, Sandwich ELISA Detection, Lateral Flow | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Monkeypox Virus E8L (RefSeq#: QNP13700.1; Met1-Thr275) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title