missing translation for 'onlineSavingsMsg'
Learn More

Monomethyl auristatin E, MedChemExpress

Product Code. 16471756
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471756 500 mg 500mg
16466338 10 mg 10mg
16428927 5 mg 5mg
16406656 100 mg 100mg
16402457 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16471756 Supplier Medchem Express Supplier No. HY15162
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Monomethyl auristatin E (MMAE; SGD-1010) is a synthetic derivative of dolastatin 10 and functions as a potent mitotic inhibitor by inhibiting tubulin polymerization. MMAE is widely used as a cytotoxic component of antibody-drug conjugates (ADCs) to treat several different cancer types.

Monomethyl auristatin E (MMAE

Specifications

Chemical Name or Material Monomethyl auristatin E
Physical Form Powder
CAS 474645-27-7
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C39H67N5O7
Synonym MMAE SGD-1010 Vedotin
Solubility Information Ethanol : 50 mg/mL (69.64 mM; Need ultrasonic) ∣DMSO : ≥ 48 mg/mL (66.85 mM)
SMILES CC(C)[C@H](NC)C(N[C@H](C(N([C@@H]([C@@H](C)CC)[C@@H](CC(N1CCC[C@@]1([H])[C@H](OC)[C@@H](C)C(N[C@H](C)[C@H](C2=CC=CC=C2)O)=O)=O)OC)C)=O)C(C)C)=O
Molecular Weight (g/mol) 717.98
Formula Weight 717.98
Percent Purity 97.54%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.