Learn More
Monomethyl auristatin E (MMAE) Antibody (08), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-31974-20ug
 View more versions of this product
Product Code. 30115289
  • 214.00 € / 20µg
Description
Monomethyl auristatin E (MMAE) Monoclonal antibody specifically detects Monomethyl auristatin E (MMAE) in Nonspecies specific samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monomethyl auristatin E (MMAE)
Monoclonal
Unconjugated
0.2 um filtered solution in PBS
Mouse
Protein A purified
RUO
Non-species specific
Purified
ELISA
08
ELISA 1:1000-1:2000
474645-27-7, Monomethyl auristatin E (GMP), vedotin
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with Monomethyl auristatin E (MMAE) conjugated to BSA
20 μg
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1