missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monosultap, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
| Packaging | Glass Bottle |
| CAS | 29547-00-0 |
| Physical Form | Crystalline |
| Industry Type | Acaricides and Insecticides |
| Quantity | 100 mg |
| Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction