missing translation for 'onlineSavingsMsg'
Learn More

Monoxerutin, TRC

Product Code. 30477746
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30477746 50 mg 50mg
30480526 100 mg 100mg
30464338 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477746 Supplier Toronto Research Chemicals Supplier No. TRCM23800050MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23869-24-1
Molecular Formula C29H34O17
Molecular Weight (g/mol) 654.57
Synonym O-(beta-Hydroxyethyl)rutin, O-(beta-Hydroxyethyl)rutoside, Venorutin 300, Verorutin 300, Z 12007, Zyma SA, 7-O-(beta-Hydroxyethyl)rutin, 7-O-(beta-Hydroxyethyl)rutoside, Mono-O-(beta-hydroxyethyl)rutoside, 2-(3,4-Dihydroxyphenyl)-5-hydroxy-7-(2-hydroxyethoxy)-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one
IUPAC Name 2-(3,4-dihydroxyphenyl)-5-hydroxy-7-(2-hydroxyethoxy)-3-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-[[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxymethyl]oxan-2-yl]oxychromen-4-one
SMILES C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(OCCO)cc(O)c4C3=O)c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C29H34O17/c1-10-19(34)22(37)24(39)28(43-10)42-9-17-20(35)23(38)25(40)29(45-17)46-27-21(36)18-15(33)7-12(41-5-4-30)8-16(18)44-26(27)11-2-3-13(31)14(32)6-11/h2-3,6-8,10,17,19-20,22-25,28-35,37-40H,4-5,9H2,1H3/t10-,17+,19-,20+,22+,23-,24+,25+,28+,29-/m0/s1
Formula Weight 654.1796
Chemical Name or Material Monoxerutin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.