missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Montana Pesticide Mixture 270 10 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Abamectin∣Acequinocyl∣Bifenazate∣Bifenthrin∣Chlormequat chloride∣Cyfluthrin∣Daminozide∣Etoxazole∣Fenoxycarb∣Imazalil∣Imidacloprid∣Myclobutanil∣Paclobutrazol∣Pyrethrins∣Spinosad∣Spirotetramat∣Trifloxystrobin |
| CAS | 71751-41-2, 57960-19-7, 149877-41-8, 82657-04-3, 999-81-5, 68359-37-5 |
| Analyte or Component Names | Abamectin∣Acequinocyl∣Bifenazate∣Bifenthrin∣Chlormequat chloride∣Cyfluthrin∣Daminozide∣Etoxazole∣Fenoxycarb∣Imazalil∣Imidacloprid∣Myclobutanil∣Paclobutrazol∣Pyrethrins∣Spinosad∣Spirotetramat∣Trifloxystrobin |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetonitrile |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Abamectin 10 μg/mL∣Acequinocyl 10 μg/mL∣Bifenazate 10 μg/mL∣Bifenthrin 10 μg/mL∣Chlormequat chloride 10 μg/mL∣Cyfluthrin 10 μg/mL∣Daminozide 10 μg/mL∣Etoxazole 10 μg/mL∣Fenoxycarb 10 μg/mL∣Imazalil 10 μg/mL∣Imidacloprid 10 μg/mL∣Myclobutanil 10 μg/mL∣Paclobutrazol 10 μg/mL∣Pyrethrins 10 μg/mL∣Spinosad 10 μg/mL∣Spirotetramat 10 μg/mL∣Trifloxystrobin 10 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?