missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 1.0M buffer soln., pH 5.5, Thermo Scientific™

Product Code. 15434669
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15434669 250 mL 250mL
15444669 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15434669 Supplier Thermo Scientific Alfa Aesar Supplier No. J62261.AK
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Format
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a morpholino propanesulfonic acid buffer.

Solubility
Soluble in water.

Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Especificaciones

Color Colorless
Concentration 1.0 M
pH 5.5
Physical Form Liquid
Quantity 250 mL
Odor Odorless
Solubility Information Soluble in water.
Chemical Name or Material MOPS buffer soln.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.