missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 101%, MP Biomedicals™

Product Code. 11496071 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496071 25 g 25g
11406081 100 g 100g
11416081 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11496071 Supplier MP Biomedicals Supplier No. 0219067025
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 71119-22-7
Molecular Formula C7H14NNaO4S
Molecular Weight (g/mol) 231.242
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
Synonym 3-[N-Morpholino]propanesulfonic acid
PubChem CID 3859613
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate
SMILES C1COCCN1CCCS(=O)(=O)[O-].[Na+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 1.01
Formula Weight 231.2
Chemical Name or Material MOPS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.