missing translation for 'onlineSavingsMsg'
Learn More

MOPS, ≥98.5% (Dry Basis), Ultrapure

Product Code. 15845178 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15845178 100 g Plastic bottle 100g
15885318 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15845178 Supplier Thermo Scientific Supplier No. J19256.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-[N-Morpholino]propane sulfonic Acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES C1COCCN1CCCS(=O)(=O)O

Specifications

Melting Point 277°C to 280°C (decomposition)
Physical Form Crystalline Powder
Quantity 100 g
Flash Point 110°C
Linear Formula HO3S(CH2)3(C4H8NO)
Packaging Plastic bottle
Beilstein 1106776
Formula Weight 209.26
Percent Purity ≥98.5%
Assay Dry basis
Chemical Name or Material MOPS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.