missing translation for 'onlineSavingsMsg'
Learn More

MOPS, 99%

Product Code. 11419733
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419733 500 g 500g
11499723 25 g 25g
11409733 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419733 Supplier Thermo Scientific Alfa Aesar Supplier No. A12914.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is mainly used in biological experiments as a buffer solution. It is also used in protein purification. Further, it is utilized for polyacrylamide gel electrophoresis.

Solubility
Soluble in water.    

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 4-Morpholinepropanesulfonic acid; 3-(4-Morpholinyl)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Melting Point 277°C to 280°C (decomposition)
Quantity 500 g
Flash Point 110°C (230°F)
Assay Percent Range 99%
Beilstein 1106776
Merck Index 14,6265
Solubility Information Soluble in water.
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
Formula Weight 209.27
Percent Purity 99%
Chemical Name or Material MOPS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.