missing translation for 'onlineSavingsMsg'
Learn More

MOPS (Fine White Crystals/Molecular Biology), Fisher BioReagents™

Product Code. 10234673
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10234673 100 g Glass Bottle 100g
10234723 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10234673 Supplier Fisher BioReagents Supplier No. BP308100
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Commonly used buffering agent

This biological buffer has a usable pH range of 6.5 to 7.9.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(4-Morpholino)propane sulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Melting Point 277°C
Color White
Decomposition Information >277°C
pH 2.5 to 4.0
Physical Form Powder or Solid
Quantity 100 g
DNase DNase free
Assay Percent Range ≥97 %
Identification Pass Test
Linear Formula CH2CH2OCH2CH2N(CH2)3SO3H
Packaging Glass Bottle
Protease Protease free
Merck Index 15, 6352
Formula Weight 209.26
Percent Purity ≥97%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material MOPS
Show More Show Less
compliance-icons
Product Identifier
  • MOPS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.