missing translation for 'onlineSavingsMsg'
Learn More

MOPS, free acid, 100.2%, MP Biomedicals™

Product Code. 11426591 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426591 500 g 500g
11406591 25 g 25g
11416591 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11426591 Supplier MP Biomedicals Supplier No. 0219483790
  • ONLINE EXCLUSIVE PRICE
    477.00 €
    451.50 € / 500g
    Save 25.50 €
    5% Off
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-[N-Morpholino]propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

For Use With (Application) Molecular Biology
Physical Form Powder
Quantity 500 g
Solution Type Buffer
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
Formula Weight 209.3
Percent Purity 1.002
Grade Molecular Biology
Chemical Name or Material MOPS
compliance-icons
Product Identifier
  • MOPS, molecular biology reagent
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: MOPS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.