missing translation for 'onlineSavingsMsg'
Learn More

MOPS, MP Biomedicals™

Product Code. 11493610 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
1 kg
Unit Size:
100g
10g
1kg
250g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493610 250 g 250g
11473610 10 g 10g
11483610 100 g 100g
11403620 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11493610 Supplier MP Biomedicals Supplier No. 0210237083
  • ONLINE EXCLUSIVE PRICE
    309.00 €
    292.95 € / 250g
    Save 16.05 €
    5% Off
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-morpholino)propanesulfonic acid, 4-morpholinepropanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 100.4%
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
Formula Weight 209.3
Chemical Name or Material MOPS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.