missing translation for 'onlineSavingsMsg'
Learn More

MOPS sodium salt, 98%

Product Code. 10651643 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10651643 25 g Glass bottle 25g
10108003 100 g Glass bottle 100g
10391284 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10651643 Supplier Thermo Scientific Acros Supplier No. 352590250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71119-22-7 | C7H14NNaO4S | 231.242 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C7H14NNaO4S
Molecular Weight (g/mol) 231.242
MDL Number MFCD00064350
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
Synonym 3-(4-Morpholino)propanesulfonic acid sodium salt
PubChem CID 3859613
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate
SMILES C1COCCN1CCCS(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 231.25
Percent Purity 98%
Chemical Name or Material MOPS sodium salt, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.