missing translation for 'onlineSavingsMsg'
Learn More

MOPS Sodium Salt Crystalline MP Biomedicals

Product Code. 11426081 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11426081 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11426081 Supplier MP Biomedicals Supplier No. 0219067091
  • ONLINE EXCLUSIVE PRICE
    1030.00 €
    972.30 € / 1kg
    Save 57.70 €
    5% Off
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 71119-22-7
Physical Form Crystalline
Quantity 1 kg
Molecular Formula C7H14NNaO4S
Synonym 3-[N-Morpholino]propanesulfonic acid
InChI Key MWEMXEWFLIDTSJ-UHFFFAOYSA-M
SMILES C1COCCN1CCCS(=O)(=O)[O-].[Na+]
IUPAC Name sodium;3-morpholin-4-ylpropane-1-sulfonate
Molecular Weight (g/mol) 231.242
PubChem CID 3859613
Formula Weight 231.2
Chemical Name or Material MOPS Sodium Salt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.