missing translation for 'onlineSavingsMsg'
Learn More

MOPSO, 99.5%, MP Biomedicals™

Product Code. 11422732 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g

Product Code. 11422732

Brand: MP Biomedicals 0215245580

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 79803-73-9
Color White
Physical Form Powder
Quantity 100 g
Molecular Formula C7H14NNaO5S
Synonym sodium 2-hydroxy-3-morpholinopropane-1-sulfonate, mopso sodium salt, 3-morpholino-2-hydroxypropanesulfonic acid sodium salt, 3-n-morpholinyl-2-hydroxypropanesulfonic acid sodium salt, sodium 2-hydroxy-3-morpholin-4-yl propane-1-sulfonate, mopso sodium salt titration, mopso, 0.2m buffer solution, ph 6.0, mopso, 0.2m buffer solution, ph 6.5, mopso, 0.2m buffer solution, ph 7.0, mopso, 0.2m buffer solution, ph 7.5
InChI Key WSFQLUVWDKCYSW-UHFFFAOYSA-M
IUPAC Name sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate
Molecular Weight (g/mol) 247.241
Formula Weight 247.2
Chemical Name or Material MOPSO
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.