missing translation for 'onlineSavingsMsg'
Learn More

Morin hydrate

Product Code. 10589643 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10589643 5 g Glass bottle 5g
10633732 10 g Glass bottle 10g
10474575 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10589643 Supplier Thermo Scientific Acros Supplier No. 354000050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 654055-01-3 | C15H10O7 | 302.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 654055-01-3 , 480-16-0
Molecular Formula C15H10O7
Molecular Weight (g/mol) 302.24
MDL Number MFCD00217054
InChI Key YXOLAZRVSSWPPT-UHFFFAOYSA-N
Synonym morin hydrate, morinhydrate, bois d,arc hydrate, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one hydrate, morin hydrate, powder, morin flavonol, morin hydrate aurantica, 2-2,4-dihydroxyphenyl-3,5,7-trihydroxy-chromen-4-one hydrate, 2 inverted exclamation marka,3,4 inverted exclamation marka,5,7-pentahydroxyflavone
PubChem CID 16219651
IUPAC Name 2-(2,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;hydrate
SMILES OC1=CC(O)=C(C=C1)C1=C(O)C(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point 300.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Beilstein 18, 239
Merck Index 14, 6269
Solubility Information Solubility in water: insoluble. Other solubilities: slightly soluble in boiling water, soluble in organic solvents
Formula Weight 302.24
Physical Form Powder
Chemical Name or Material Morin hydrate
Show More Show Less
compliance-icons
Produktbeteckning
  • Morin hydrate
Riskkategorier
  • LONG-TERM AQUATIC HAZARD Chronic 2
Faroangivelser
  • H411-Toxic to aquatic life with long lasting effects.
Skyddsangivelse
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.