missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Angiopoietin-3 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB738
This item is not returnable.
View return policy
Description
Angiopoietin-3 Monoclonal specifically detects Angiopoietin-3 in Mouse samples. It is validated for Western Blot.
Specifications
| Angiopoietin-3 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Angpt4 | |
| E. coli-derived recombinant mouse Angiopoietin-3 | |
| 500 μg | |
| Primary | |
| Detects mouse Angiopoietin-3 in direct ELISA and Western blots. In Western blots, less than 1% cross-reactivity with recombinant human (rh) Angiopoietin-1, rhAngiopoietin-2, rhAngiopoietin-like factor, and rhAngiopoietin-4 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 113504 | |
| Western Blot 1 ug/mL | |
| ANG3, ANG4, ANGPT3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 51378 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction