missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ALCAM/CD166 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
208.00 € - 1720.00 €
Specifications
| Antigen | ALCAM/CD166 |
|---|---|
| Clone | 105902 |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Western Blot, Flow Cytometry, CyTOF, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18759497
![]() |
R&D Systems
MAB6561-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18736249
![]() |
R&D Systems
MAB6561-100 |
100 μg |
428.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18755519
![]() |
R&D Systems
MAB6561-500 |
500 μg |
1720.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALCAM/CD166 Monoclonal specifically detects ALCAM/CD166 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready, ELISA Capture (Matched Antibody Pair).Specifications
| ALCAM/CD166 | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q13740 | |
| 214 | |
| Mouse myeloma cell line NS0-derived recombinant human ALCAM/CD166 Trp28-Ala526 Accession # Q13740 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ALCAM/CD166 in Western blots. Shows approximately 50% cross-reactivity with recombinant mouse OCAM and no cross-reactivity with recombinant human (rh) BCAM, rhEpCAM, rhMCAM, or rhNCAM-L1. |
| 105902 | |
| Western Blot, Flow Cytometry, CyTOF, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activated leucocyte cell adhesion molecule, activated leukocyte cell adhesion moleculeFLJ38514, CD166, CD166 antigen, MEMDMGC71733 | |
| ALCAM | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title