Learn More
CD326 Mouse anti-Human, Brilliant Violet 786, Clone: EBA-1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565685
Description
The EBA-1 monoclonal antibody specifically binds to human CD326. CD326 is an approximately 40 kDa type 1 transmembrane glycoprotein and adhesion molecule that mediates intercellular adhesive interactions. CD326 is also known as epithelial adhesion molecule (EpCAM), epithelial glycoprotein 2 (EGP-2), and epithelial surface antigen (ESA). The epithelial cells present in non-squamous epithelia and tumors derived from such cells show EpCAM expression. The normal epithelial cells reactive with anti-EpCAM antibodies are those present in the (lower) respiratory tract; the (lower) gastrointestinal tract; tubules in the kidney; the surface epithelium of the ovary; the exocrine and endocrine pancreas; secondary germ cells of telogenic hair follicles; and secretory tubules of sweat glands in the skin, whereas the epidermis is negative. In addition, all epithelial cells in the thyroid and epithelial cells in the thymus show EpCAM expression, while the outer cortex and Hassall’s corpuscles have low expression. In the liver, only the bile ducts appear to be positive with anti-EpCAM antibodies. Non-squamous- carcinoma cells have high EpCAM expression; some squamous carcinoma cells. Tumors arising from non-epithelial cells, such as lymphoma, mesothelioma, neuroblastoma, and melanoma, do not express EpCAM.
The antibody was conjugated to BD Horizon BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
| CD326 | |
| Monoclonal | |
| Brilliant Violet 786 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Breast carcinoma-associated mucin BCA-225 | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| EBA-1 | |
| EPCAM; EGP; ESA; GA733-2; hEGP-2; KSA; M4S1; MIC18; MK-1; TACSTD1; TROP1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 λ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.