missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG (H+L) Secondary Antibody (4D2D9G8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-51523SS
This item is not returnable.
View return policy
Descripción
IgG (H+L) Monoclonal antibody specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA.
Especificaciones
| IgG (H+L) | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
| IGHG1 | |
| Human IgG was isolated from human sera and purified by chromatography. | |
| 0.025 mL | |
| Secondary | |
| Monoclonal anti-Human IgG is derived from a hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. It is specific for the whole human IgG molecule, and has no cross reactivity with human IgM molecule as determined by an ELISA. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 4D2D9G8 | |
| Unconjugated | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3500 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido