Learn More
Streptococcus agalactiae, Mouse, Clone: Strep12-424.1, Abnova™
Mouse monoclonal antibody raised against Streptococcus agalactiae.
Brand:  Abnova MAB8387.100ug
Product Code. 16087346
  • 388.00 € / 100µg
Specifications
Streptococcus agalactiae
Monoclonal
Unconjugated
The optimal working dilution should be determined by the end user.
Mouse
100 μg
Primary
Bacteria
Liquid
ELISA
Strep12-424.1
Mouse monoclonal antibody raised against Streptococcus agalactiae.
In PBS, pH 7.4
Native Streptococcus agalactiae cells isolated from mastitic milk.
RUO
These antibodies recognize Strep. agalactiae and cross-react weakly with Strep. dysgalactiae. They do not crossreact with Strep. uberis, Strep. bovis, Strep. faecalis, Staph. aureus, Mycoplasma bovis, Mycoplasma californicum, Mycoplasma bovigenitalium, E. coli, or Corynebacterium bovis.
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
IgM