missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 12 publications
226.00 € - 2060.00 €
Specifications
| Antigen | IL-10 |
|---|---|
| Clone | 25209 |
| Dilution | Western Blot 5 ug/mL, Neutralization 0.005-0.03 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18780978
![]() |
R&D Systems
MAB2171-SP |
25 μg |
226.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18737499
![]() |
R&D Systems
MAB2171-100 |
100 μg |
527.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18754469
![]() |
R&D Systems
MAB2171-500 |
500 μg |
2060.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-10 Monoclonal specifically detects IL-10 in Human samples. It is validated for Western Blot, Neutralization.Specifications
| IL-10 | |
| Western Blot 5 ug/mL, Neutralization 0.005-0.03 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P22301 | |
| 3586 | |
| E. coli-derived recombinant human IL-10 Ser19-Asn178 Accession # P22301 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human IL-10 in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse IL-10. |
| 25209 | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF | |
| IL10 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title