Learn More
Plasma Cell, Mouse, Clone: LIV3G11, Abnova™
Mouse monoclonal antibody raised against Plasma Cell.
Brand:  Abnova MAB11268.100ug
Product Code. 16050676
  • 503.00 € / 100µg
Specifications
Plasma Cell
Monoclonal
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) The optimal working dilution should be determined by the end user.
Mouse
Protein G purification
RUO
Human
Liquid
Immunohistochemistry (PFA fixed)
LIV3G11
Mouse monoclonal antibody raised against Plasma Cell.
In PBS (0.05% BSA, 0.05% sodium azide)
Pancreatic cancer related serum mucin isolated from human.
100 μg
Primary
Store at 4°C.
IgG2a