missing translation for 'onlineSavingsMsg'
Learn More

PPP1R1C, Mouse, Clone: 2G7, Abnova™

Product Code. 16198653
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
200µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16198653 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16198653 Supplier Abnova Supplier No. H00151242M01A.200uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against a full-length recombinant PPP1R1C.

Sequence: MEPNSPKKIQFAVPVFQSQIAPEAAEQIRKRRPTPASLVILNEHNPPEIDDKRGPNTQGELQNASPKQRKQSVYTPPTIKGVKHLKGQNESAFPEEEEGTNEREEQRDH

Specifications

Antigen PPP1R1C
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant PPP1R1C.
Formulation ascites with no preservative
Gene PPP1R1C
Gene Accession No. BC017943.1
Gene Alias IPP5
Gene Symbols PPP1R1C
Host Species Mouse
Immunogen PPP1R1C (AAH17943.1,1 a.a. ∽ 109 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 151242
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.