missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rat IgG2a Heavy Chain Secondary Antibody (MARG2a-1), Novus Biologicals™

Product Code. 18141964 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
250 μg
Unit Size:
250µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18141964 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18141964 Supplier Novus Biologicals Supplier No. NBP268423250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

IgG2a Heavy Chain Monoclonal specifically detects IgG2a Heavy Chain in Rat samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG2a Heavy Chain
Applications ELISA
Classification Monoclonal
Clone MARG2a-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution ELISA
Formulation PBS
Host Species Mouse
Immunogen Purified rat IgG2a
Quantity 250 μg
Test Specificity Mouse anti Human beta-2 Adrenergic Receptor antibody, clone R11E1, recognizes human beta 2 adrenergic receptor (beta2AR), a member of the G protein-coupled receptor superfamily.
Target Species Rat
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.