missing translation for 'onlineSavingsMsg'
Learn More

Mouse Arylsulfatase G/ARSG Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18012612
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18012612 100 μg 100µg
18748557 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18012612 Supplier R&D Systems Supplier No. AF4600

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Arylsulfatase G/ARSG Polyclonal specifically detects Arylsulfatase G/ARSG in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arylsulfatase G/ARSG
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. Q3TYD4
Gene Alias ARSG, arylsulfatase G, ASG, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.4, KIAA1001ASG
Gene Symbols ARSG
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse Arylsulfatase G/ARSG Gly17-Val525 Accession # Q3TYD4
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22901
Test Specificity Detects mouse Arylsulfatase G/ARSG in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant mouse ARSA, recombinant human (rh) ARSB and rhARSJ is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.