missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Axl Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
305.00 € - 537.00 €
Specifications
| Antigen | Axl |
|---|---|
| Clone | 175128 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.2 ug/10^6 cells |
| Applications | Flow Cytometry |
Description
Axl Monoclonal specifically detects Axl in Mouse samples. It is validated for Flow Cytometry.Specifications
| Axl | |
| 0.2 mg/mL | |
| Flow Cytometry | |
| Alexa Fluor 647 | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| AI323647, ARK, AXL oncogene, AXL receptor tyrosine kinase, AXL transforming sequence/gene, EC 2.7.10, EC 2.7.10.1, JTK11, Tyro7, tyrosine-protein kinase receptor UFO, UFO, UFOoncogene AXL | |
| AXL | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse Axl in flow cytometry. |
| 175128 | |
| Flow Cytometry 0.2 ug/10^6 cells | |
| Monoclonal | |
| Rat | |
| Mouse | |
| Q00993 | |
| 558 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Axl Ala19-Trp445 Accession # Q00993 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title