missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse B7-H3 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB1397-SP
This item is not returnable.
View return policy
Description
B7-H3/CD276 Monoclonal antibody specifically detects B7-H3/CD276 in Mouse samples. It is validated for Western Blot
Specifications
| B7-H3/CD276 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| B7H3, B7H34Ig-B7-H3, B7-H3B7 homolog 3, CD276, CD276 antigen, CD276 molecule, Costimulatory molecule | |
| Mouse myeloma cell line NS0-derived mouse B7-H3, Val29-Phe244, Accession # Q8VE98.1 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot | |
| 2741D | |
| Western Blot 1 ug/mL | |
| Q8VE98.1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 80381 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction