missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse beta-NGF Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB1156-SP
This item is not returnable.
View return policy
Description
beta-NGF Monoclonal specifically detects beta-NGF in Mouse samples. It is validated for ELISA.
Specifications
| beta-NGF | |
| Monoclonal | |
| Unconjugated | |
| Q6LDU8 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 4803 | |
| Mouse | |
| Purified |
| ELISA | |
| 2529F | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Beta nerve growth factor precursor, beta-nerve growth factor, betaNGF, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFB, NGFBMGC161428 | |
| Mouse myeloma cell line, NS0-derived mouse beta-NGF, Ser122-Gly241, Accession # Q6LDU8 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction