missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cadherin-17 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8524
This item is not returnable.
View return policy
Description
Cadherin-17 Monoclonal specifically detects Cadherin-17 in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Cadherin-17 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, Cadherin17, CDH16, CDH17, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 1015 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 1262C | |
| Western Blot 0.1 ug/mL, Simple Western 1 ug/mL, Immunohistochemistry 1-25 ug/mL | |
| Q9R100 | |
| Cdh17 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cadherin-17 Glu22-Met786 Accession # Q9R100 | |
| 100 μg | |
| Primary | |
| Detects mouse Cadherin-17 in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction