missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL1/I-309/TCA-3 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
209.00 € - 1960.00 €
Specifications
| Antigen | CCL1/I-309/TCA-3 |
|---|---|
| Clone | 148113 |
| Dilution | Western Blot 1 ug/mL, Neutralization 2-10 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18758019
![]() |
R&D Systems
MAB845-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18716108
![]() |
R&D Systems
MAB845-100 |
100 μg |
496.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18701407
![]() |
R&D Systems
MAB845-500 |
500 μg |
1960.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL1/I-309/TCA-3 Monoclonal specifically detects CCL1/I-309/TCA-3 in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
| CCL1/I-309/TCA-3 | |
| Western Blot 1 ug/mL, Neutralization 2-10 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| P10146 | |
| 6346 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CCL1/I-309/TCA-3 Lys24-Cys92 Accession # P10146 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CCL1 in direct ELISAs and Western blots. In direct ELISAs, does not cross-react with recombinant cotton rat CCL3, 4, 5, recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 23, 24, 25, 26, 27, 28, recombinant mouse CCL2, 3, 4,56, 7, 8, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, 28, or recombinant rat CCL20. |
| 148113 | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine 1, chemokine (C-C motif) ligand 1, I-309, inflammatory cytokine I-309, P500, SCYA1, SCYA1Small-inducible cytokine A1, SCYA2, SISe, small inducible cytokine A1 (I-309, homologous to mouse Tca-3), T lymphocyte-secreted protein I-309, TCA3, TCA-3 | |
| CCL1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title