missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL2/JE/MCP-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 16 publications
Brand: R&D Systems MAB479-100
This item is not returnable.
View return policy
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Neutralization.
Specifications
| CCL2/JE/MCP-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6347 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Neutralization | |
| 123616 | |
| Neutralization 0.75-3.0 ug/mL | |
| P10148 | |
| Ccl2 | |
| E. coli-derived recombinant mouse CCL2/JE/MCP-1 Gln24-Arg96 Accession # P10148 | |
| 100 μg | |
| Primary | |
| Detects mouse CCL2/JE/MCP-1 in ELISAs. Does not cross-react with recombinant human (rh) CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 27, 28, rmCCL1, 3, 4, 5, 6, 7, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, 28, or rrCCL20. | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction