Learn More
Mouse CD117/c-kit PE-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand:  R&D Systems FAB1356P-100
 View more versions of this product
Product Code. 18770148
  • 289.00 € / 100 tests
Description
CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD117/c-kit
Monoclonal
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, KIT, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCF R, SCFR, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein
Rat
Protein A or G purified from hybridoma culture supernatant
Primary
Detects mouse SCF R in direct ELISAs.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG2a
Flow Cytometry
180627
Flow Cytometry 10 μL/106 cells
P05532
KIT
Mouse myeloma cell line NS0-derived recombinant mouse SCF R, Gln25-Thr519 (Ala207Glu), Accession No. P05532
100 Tests
3815.0
Mouse
Supernatant