missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD133 Antibody, R&D Systems™

Rat Monoclonal Antibody
209.00 € - 486.00 €
Specifications
| Antigen | CD133 |
|---|---|
| Clone | 217106 |
| Dilution | Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 5-25 μg;/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18782667
![]() |
R&D Systems
MAB11332-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18703997
![]() |
R&D Systems
MAB11332-100 |
100 μg |
486.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD133 Monoclonal specifically detects CD133 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
| CD133 | |
| Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 5-25 μg;/mL, CyTOF-ready | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| O54990 | |
| 8842.0 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 217106 | |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AC133, AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, PROM1, prominin (mouse)-like 1, prominin 1, PROML1 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD133, Accession No. O54990 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CD133 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title