missing translation for 'onlineSavingsMsg'
Learn More

Mouse CD30/TNFRSF8 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18793358
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18793358 25 μg 25µg
18121091 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18793358 Supplier R&D Systems Supplier No. MAB8521SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

CD30/TNFRSF8 Monoclonal specifically detects CD30/TNFRSF8 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD30/TNFRSF8
Applications Western Blot, ELISA
Classification Monoclonal
Clone 115705
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q60846
Gene Alias CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8
Gene Symbols TNFRSF8
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD30/TNFRSF8 Phe19-Thr281 Accession # Q60846
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 943
Test Specificity Detects mouse CD30/TNFRSF8 in ELISAs and Western blots. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse (rm) TNF RI/TNFRSF1A, recombinant human (rh) TNF RI/TNFRSF1A, rhTNF RII/TNFRSF1B, rmTNF-alpha/TNFSF2, rhTNF-alpha/TNFSF2, rhTNF-beta/TNFSF1, rhCD30/TNFRSF8, or rmCD30 Ligand/TNFSF8 is observed. In Western blots, no cross-reactivity with rhNGFR/TNFRSF16, rmOPG/TNFRSF11B, rmRANK/TNFRSF11A, rmTNF RI/TNFRSF1A, rmTNF RII/TNFRSF1B, rmEDAR, rmTAJ/TNFSF19, rmLTR beta/TNFRSF3, rm4-1BB/TNFRSF9, rmCD27/TNFRSF7, rhCD30/TNFRSF8, rmCD40/TNFRSF5, rhDR3/TNFRSF12, rhDR6/TNFRSF21, rmFas/TNFRSF6, rmGITR/TNFRSF18, or rhHVEM/TNFRSF14 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.