missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD30/TNFRSF8 Antibody, R&D Systems™

Description
CD30/TNFRSF8 Monoclonal specifically detects CD30/TNFRSF8 in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
| Antigen | CD30/TNFRSF8 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 115705 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q60846 |
| Gene Alias | CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 |
| Gene Symbols | TNFRSF8 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?