missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD39L3/ENTPD3 Antibody, R&D Systems™

Sheep Polyclonal Antibody
214.00 € - 489.00 €
Specifications
| Antigen | CD39L3/ENTPD3 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18778757
![]() |
R&D Systems
AF4464-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18088781
![]() |
R&D Systems
AF4464 |
100 μg |
489.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD39L3/ENTPD3 Polyclonal specifically detects CD39L3/ENTPD3 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| CD39L3/ENTPD3 | |
| Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Sheep | |
| Mouse | |
| ATPDase, CD39 antigen-like 3, CD39L3, CD39L3EC 3.6.1.5, CD39-like 3, EC 3.6.1, Ecto-apyrase 3, Ecto-ATP diphosphohydrolase 3, Ecto-ATPase 3, Ecto-ATPDase 3, ectonucleoside triphosphate diphosphohydrolase 3, ENTPD3, FLJ93839, HB6, HB6NTPDase 3, NTPDase-3 | |
| ENTPD3 | |
| IgG | |
| Antigen Affinity-purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| AAH79871 | |
| 956 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse CD39L3/ENTPD3 Gln44-Pro485 Accession # AAH79871 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse CD39L3/ENTPD3 in direct ELISAs and Western blots. In direct ELISAs, approximately 30% cross-reactivity with recombinant human CD39L3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title