missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD47 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB18661-100
This item is not returnable.
View return policy
Description
CD47 Monoclonal specifically detects CD47 in Mouse samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
Specifications
| CD47 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAP, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen | |
| Mouse myeloma cell line NS0-derived mouse CD47, Gln19-Pro158, Accession # NP_034711 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG1 |
| Flow Cytometry, Blocking Assay, CyTOF | |
| 974222 | |
| Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 0.08-0.8 ug/mL, CyTOF-ready | |
| NP_034711 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 961 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction