missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CRISP-1 Antibody, R&D Systems™

Rat Monoclonal Antibody
184.00 € - 377.00 €
Specifications
| Antigen | CRISP-1 |
|---|---|
| Clone | 840017 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 10-20 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18789138
![]() |
R&D Systems
MAB4675-SP |
25 μg |
184.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18705569
![]() |
R&D Systems
MAB4675 |
100 μg |
377.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CRISP-1 Monoclonal specifically detects CRISP-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
| CRISP-1 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 10-20 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acidic epididymal glycoprotein homolog, acidic epididymal glycoprotein-like 1, AEGL1, AEG-related protein, ARPAEG-like protein, CRISP1, CRISP-1AEGL1HSCRISP1D, cysteine-rich secretory protein 1, cysteine-rich secretory protein-1 delta, HSCRISP1G, HUMARP | |
| Crisp1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 840017 | |
| Western Blot, Immunohistochemistry, Neutralization | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q03401 | |
| 167 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CRISP-1 Gln20-His244 Accession # Q03401 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CRISP-1 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title