Learn More
Mouse CXCR2/IL-8RB PE-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 22 publications
Brand:  R&D Systems FAB2164P-025
 View more versions of this product
Product Code. 18756677
  • 237.00 € / 25 tests
Description
CXCR2/IL-8RB Monoclonal specifically detects CXCR2/IL-8RB in Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCR2/IL-8RB
Monoclonal
R-PE
P35343
CXCR2
C6 rat glioma cell line transfected with mouse CXCR2 Met1-Leu359 Accession # P35343
25 Tests
3579
Mouse
Purified
Flow Cytometry
242216
Flow Cytometry 10 uL/10^6 cells
CD182, CD182 antigen, CDw128b, chemokine (C-X-C motif) receptor 2, chemokine (CXC) receptor 2, CMKAR2, C-X-C chemokine receptor type 2, CXC-R2, CXCR-2, CXCR2 gene for IL8 receptor type B, GRO/MGSA receptor, High affinity interleukin-8 receptor B, IL-8 RB, IL-8 receptor type 2, IL-8R B, IL8R2, IL8RA, IL8RB, interleukin 8 receptor B, interleukin 8 receptor type 2, interleukin-8 receptor type B
Rat
Protein A or G purified from hybridoma culture supernatant
Primary
Stains mouse CXCR2-transfected cells but not CXCR1 transfectants.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2a