missing translation for 'onlineSavingsMsg'
Learn More

Mouse Cystatin E/M Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18782808
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18782808 25 μg 25µg
18199730 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782808 Supplier R&D Systems Supplier No. MAB1284SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Cystatin E/M/CST6 Monoclonal specifically detects Cystatin E/M/CST6 in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cystatin E/M/CST6
Applications Western Blot, Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 216820
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 7 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_082899
Gene Alias CST6, cystatin 6, cystatin E/M, cystatin M, cystatin M/E, Cystatin-6, Cystatin-E, cystatin-M, cysteine proteinase inhibitor
Gene Symbols CST6
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Cystatin E/M Glu29-Val149 Accession # NP_082899
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1474
Test Specificity Detects mouse Cystatin E/M in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Cystatin B, S, E/M, SA, S/N, recombinant mouse (rm) Cystatin A, C, D, rhFetuin A, B, rmHPRG, or rhKininogen is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.