missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Dkk-3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB948-500
This item is not returnable.
View return policy
Description
Dkk-3 Monoclonal specifically detects Dkk-3 in Mouse samples. It is validated for Western Blot.
Specifications
| Dkk-3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9QUN9 | |
| DKK3 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Dkk-3 Ala22-Ile349 Accession # Q9QUN9 | |
| 500 μg | |
| Primary | |
| Detects mouse Dkk-3 in direct ELISAs and Western blots. Does not cross-react with recombinant mouse Dkk-1, recombinant human (rh) Dkk-3, or rhDkk-4. | |
| Mouse | |
| Purified |
| Western Blot | |
| 153103 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| dickkopf (Xenopus laevis) homolog 3, dickkopf 3, dickkopf homolog 3 (Xenopus laevis), Dickkopf-3, dickkopf-related protein 3, Dkk3, Dkk-3, hDkk-3, regulated in glioma, REIC, REICdkk-3, RIG, RIG-like 5-6, RIG-like 7-1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27122 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction