missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse EMMPRIN/CD147 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB7721
This item is not returnable.
View return policy
Description
EMMPRIN/CD147 Monoclonal specifically detects EMMPRIN/CD147 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| EMMPRIN/CD147 | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| 5F7, basigin, basigin (Ok blood group), BSG, CD147, CD147 antigen, Collagenase stimulatory factor, EMMPRINTCSF, Extracellular matrix metalloproteinase inducer, Leukocyte activation antigen M6, M6, OK, OK blood group antigen, Tumor cell-derived collagenase stimulatory factor | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 682 | |
| Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 1159A | |
| Western Blot 1:1000 dilution, Immunohistochemistry 1:200-1:400 dilution | |
| P18572 | |
| BSG | |
| NS0 mouse myeloma cell line transfected with mouse EMMPRIN/CD147 Ala22-Pro209 Accession # P18572 | |
| 100 μg | |
| Primary | |
| Detects mouse EMMPRIN/CD147 in direct ELISAs. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction