missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Epimorphin/Syntaxin 2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2568-SP
This item is not returnable.
View return policy
Description
Epimorphin/Syntaxin 2 Polyclonal specifically detects Epimorphin/Syntaxin 2 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Epimorphin/Syntaxin 2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EPIMepimorphin, EPM, STX2, STX2AMGC51014, STX2Bsyntaxin-2, STX2CEpimorphin, Syntaxin 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2054 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q00262 | |
| STX2 | |
| E. coli-derived recombinant mouse Epimorphin/Syntaxin 2 Met1-Arg189 Accession # Q00262 | |
| 25 μg | |
| Primary | |
| Detects mouse Epimorphin/Syntaxin 2 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human Epimorphin is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction